Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENO4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549055

Catalog No. PA549055

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be required for sperm motility and function.
Specifications

Specifications

ENO4
Polyclonal
PBS with 0.09% sodium azide
A6NNW6, Q8C042
Enolase-like protein ENO4, ENO4, C10orf134
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
226265, 387712
Western Blot
Unconjugated
ENO4
Liquid
ENO4
KLH conjugated synthetic peptide between 388-414 amino acids from the Central region of human ENO4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit