Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 1 Mouse anti-Human, Biotin, Clone: 186, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243701B

Catalog No. NBP243701B


Add to cart

Description

Description

Enolase 1 Monoclonal antibody specifically detects Enolase 1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Enolase 1
186
Western Blot
Purified
ENO1
Recombinant protein encompassing a sequence within the center region of human ENO1. The exact sequence is proprietary.
Protein G purified
Cancer, Core ESC Like Genes, Stem Cell Markers
Primary
2023
Western Blot
Biotin
PBS with 0.05% Sodium Azide
2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit