Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 2/Neuron-specific Enolase Mouse anti-Human, Clone: 972458, R&D Systems™

Mouse Monoclonal Antibody

$116.82 - $320.50

Specifications

Antigen Enolase 2/Neuron-specific Enolase
Clone 972458
Dilution Western Blot 0.5 ug/mL, Immunocytochemistry 1-25 ug/mL
Applications Immunocytochemistry, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB51691100 View Documents R&D Systems
MAB51691100
100 ug Each for $320.50
Add to cart
 
MAB51691SP View Documents R&D Systems
MAB51691SP
25 ug Each for $116.82
Add to cart
 
Description

Description

Enolase 2/Neuron-specific Enolase Monoclonal antibody specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications

Specifications

Enolase 2/Neuron-specific Enolase
Western Blot 0.5 ug/mL, Immunocytochemistry 1-25 ug/mL
Unconjugated
Mouse
Human
P09104
2026
IgG2a
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Enolase 2/Neuron-specific Enolase in direct ELISAs.
972458
Immunocytochemistry, Western Blot
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, ENO2, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, Neuronspecific Enolase, NSE
E. coli-derived recombinant human Enolase 2/Neuron-specific Enolase Met1-Leu434 Accession # P09104
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit