Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enolase 2/Neuron-specific Enolase Antibody (SPM347), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254590PEATT594
Description
Enolase 2/Neuron-specific Enolase Monoclonal specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Flow Cytometry.Specifications
Enolase 2/Neuron-specific Enolase | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
ENO2 | |
A synthetic peptide of human Enolase 2/Neuron-specific Enolase (around aa416-433; exact sequence is proprietary) (Uniprot: P09104) | |
0.1 mL | |
Primary | |
Human | |
IgG2b κ |
Flow Cytometry | |
SPM347 | |
Flow Cytometry | |
2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
Mouse | |
Protein A or G purified | |
Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
2026.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction