Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 2/Neuron-specific Enolase Mouse anti-Human, PE/Cy5.5, Clone: SPM347, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254590PECY55

Catalog No. N254590PEC5


Add to cart

Description

Description

Enolase 2/Neuron-specific Enolase Monoclonal antibody specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Enolase 2/Neuron-specific Enolase
SPM347
Flow Cytometry
Conjugated, Purified
ENO2
A synthetic peptide corresponding to aa416-433 of human NSE gamma (exact sequence is proprietary)
Protein A or G purified
Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary
2026.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit