Learn More
eNOS Mouse anti-Human, DyLight 405, Clone: 6H2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP151582V
Catalog No. NBP151582V
Add to cart
Description
Description
eNOS Monoclonal antibody specifically detects eNOS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
eNOS
6H2
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
NOS3
Purified recombinant fragment of human eNOS expressed in E. coli. [UniProt# P29474]
Ammonium sulfate precipitation
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII
Mouse
IgG1
0.1 ml
Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific
Primary
4846