Learn More
CEP Enpac™ Easy Berm™ Spill Containment Unit
Handles even large jobs such as leaking tanker trucks
Manufacturer:  CEP 5806BK
 View more versions of this product
Catalog No. 19-047-818
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Lightweight, portable spill containment unit
  • Unique snap-up support strips provide sturdy support for the collapsible side walls when the unit is open, while allowing easy accessibility and placement
  • Folds up to the most compact size per square foot of any comparable unit, making it a convenient component for spill response kits
  • Tough XR-5 construction holds up against rips, tears, and punctures while offering excellent resistance to the widest range of chemicals, acids, and fuels
Specifications
Specifications
Safety