Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP Enpac™ Prowler™ Spill Containers

Opaque to allow visually checks of content level

Manufacturer:  CEP 5920OP

 View more versions of this product

Catalog No. 19-047-805


Add to cart

Description

Description

  • Strongest, lightest-weight pop-up containment pool
  • Polyethylene construction with high-density polyethylene tapes and low-density coating
  • Superior chemical resistance and cold resistance up to -54°C (-65°F)
  • Excellent abrasion, puncture, and UV resistance
  • Compact packaging for easy storage
  • Color-coded by size

Specifications

Specifications

20 gal.
Opaque Blue Ring
13 in.
HDPE tapes
Polyethylene
Safety
75.7L
It offers excellent resistance to the widest range of chemicals, acids, leaded and unleaded gasoline, diesel fuels and other hazardous materials
33cm
Chemical, cold, abrasion, puncture, UV resistance
Prowler Pool
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit