Learn More
Enpac™ Prowler™ Spill Containers
Opaque to allow visually checks of content level
Manufacturer:  COMPLETE ENVIRONMENTAL PRODUCTS 5966OP
 View more versions of this product
Catalog No. 19-047-809
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
66 gal.
Opaque Red Ring
13 in.
HDPE tapes
Polyethylene
249.8L
It offers excellent resistance to the widest range of chemicals, acids, leaded and unleaded gasoline, diesel fuels and other hazardous materials
33cm
Chemical, cold, abrasion, puncture, UV resistance
Prowler Pool