Learn More
ENPP-3/CD203c Mouse anti-Human, DyLight™ 650, Clone: NP4D6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP144643C
Catalog No. NBP144643C
Add to cart
Description
Description
ENPP-3/CD203c Monoclonal specifically detects ENPP-3/CD203c in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ENPP-3/CD203c
NP4D6
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunocytochemistry
CD203c, CD203c antigen, dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), ectonucleotide pyrophosphatase/phosphodiesterase 3, ectonucleotide pyrophosphatase/phosphodiesterase family member 3, E-NPP 3, gp130RB13-6, NPP3, PD-IBETA, PDNP3B10, Phosphodiesterase I beta, Phosphodiesterase I/nucleotide pyrophosphatase 3, phosphodiesterase-I beta
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody NP4D6 reacts with CD203c, a transmembrane ectoenzyme expressed on basophils and mast cells, and overexpressed upon their activation. HLDA VIII
Flow Cytometry, Immunocytochemistry, Immunofluorescence
DyLight 650
50 mM sodium borate with 0.05% sodium azide
ENPP3
HEK-293 cells transfected with human CD203c
0.1 mL
Primary
5169
Human