Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP1 Polyclonal Antibody
ENPP1 Polyclonal Antibody
Supplier: Thermo Scientific PA572943
Description
ENPP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
ENPP1 | |
Polyclonal | |
Unconjugated | |
ENPP1 | |
4833416E15Rik; AI428932; alkaline phosphodiesterase 1; Alkaline phosphodiesterase I; ARHR2; C76301; CD203c; COLED; ectonucleotide pyrophosphatase/phosphodiesterase 1; ectonucleotide pyrophosphatase/phosphodiesterase family member 1; Ectonucleotide pyrophosphatase/phosphodiesterase family member 1, secreted form; E-NPP 1; Enpp1; E-NPP1; HGNC:3356; Ly-41; Ly-41 antigen; Lymphocyte antigen 41; M6S1; Membrane component chromosome 6 surface marker 1; membrane component, chromosome 6, surface marker 1; NPP1; NPPase; NPPS; Nucleotide diphosphatase; Nucleotide pyrophosphatase; OTTHUMP00000043194; PC1; PC-1; Pca; PCA1; Pca-1; PDNP1; Phosphodiesterase I/nucleotide pyrophosphatase 1; plasma-cell membrane glycoprotein 1; plasma-cell membrane glycoprotein PC-1; ttw; twy | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5167 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.18 mg/mL | |
PBS with 30% glycerol and 0.05% sodium azide | |
P22413 | |
ENPP1 | |
A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413]. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction