Learn More
CD203c Mouse anti-Human, PE, Clone: NP4D6, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15783
Catalog No. 50-113-7764
Add to cart
Description
Description
The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008]
Specifications
Specifications
CD203c
Monoclonal
PE
ENPP3
Liquid
ENPP3
human CD203c
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
NP4D6
PBS with 0.2% BSA and 15mM sodium azide
O14638
B10, CD203c, NPP3, PD-IBETA, PDNP3, E-NPP 3
Mouse
IgG1
100 tests
4° C
Primary
5169