Learn More
ENPP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 173051AP
Catalog No. 50-173-1499
Add to cart
Description
Description
NPP4, also known as ENPP4 (ectonucleotide pyrophosphatase/phosphodiesterase family member 4), is a 453 amino acid single-pass type I membrane protein that belongs to the nucleotide pyrophosphatase/phosphodiesterase family. The gene that encodes NPP4 consists of approximately 16, 736 bases and maps to human chromosome 6p21.1. Making up nearly 6% of the human genome, chromosome 6 contains around 1, 200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene, and Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. Notably, the PARK2 gene, which is associated with Parkinson ft.s disease, and the genes encoding the major histocompatibility complex proteins are also located on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.
Specifications
Specifications
ENPP4
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BTJ4, Q9Y6X5
E NPP 4, ENPP4, KIAA0879
Rabbit
IgG
150 μL
-20°C
Primary
224794, 22875, 301261
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ENPP4
Liquid
Enpp4
ENPP4 Fusion Protein Ag11192
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat