Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENPP7 Monoclonal Antibody (10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529243
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ENPP7.
ENPP7 converts sphingomyelin to ceramide. ENPP7 also has phospholipase C activity toward palmitoyl lyso-phosphocholine. ENPP7 does not appear to have nucleotide pyrophosphatase activity.
Specifications
ENPP7 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q6UWV6 | |
ENPP7 | |
Recombinant Human ENPP7 protein (Met1-Ser439). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry | |
10 | |
Unconjugated | |
ENPP7 | |
alkaline sphingomyelin phosphodiesterase; alkaline sphingomyelinase; Alk-SMase; ectonucleotide pyrophosphatase/phosphodiesterase 7; ectonucleotide pyrophosphatase/phosphodiesterase family member 7; E-NPP 7; ENPP7; E-NPP7; Gm254; Intestinal alkaline sphingomyelinase; NPP7; NPP-7; sphingomyelinase, intestinal alkaline; UNQ3077/PRO9912 | |
Mouse | |
Protein A | |
RUO | |
339221 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction