Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENPP7 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547705
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant human ENPP-7 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ENPP7 converts sphingomyelin to ceramide. ENPP7 also has phospholipase C activity toward palmitoyl lyso-phosphocholine. ENPP7 does not appear to have nucleotide pyrophosphatase activity.
Specifications
ENPP7 | |
Polyclonal | |
Unconjugated | |
ENPP7 | |
alkaline sphingomyelin phosphodiesterase; alkaline sphingomyelinase; Alk-SMase; ectonucleotide pyrophosphatase/phosphodiesterase 7; ectonucleotide pyrophosphatase/phosphodiesterase family member 7; E-NPP 7; ENPP7; E-NPP7; Gm254; Intestinal alkaline sphingomyelinase; NPP7; NPP-7; sphingomyelinase, intestinal alkaline; UNQ3077/PRO9912 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
238011 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q3TIW9 | |
ENPP7 | |
Mouse myeloma cell line NS0-derived recombinant mouse ENPP-7/Alk-SMase Ala22-Gln421. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction