Learn More
BioVision IncSupplier Diversity Partner Enteropeptidase/Enterokinase Cleavage Kit: Convenient tool to efficiently remove tags from recombinant fusion proteins containing accessible enteropeptidase-specific recognition sequence. Sufficient for cleaving at least 5 mg of target protein. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K760100
Enteropeptidase/Enterokinase Cleavage Kit: Enteropeptidase (Enterokinase, EC 3.4.21.9) is a serine protease involved in activation of trypsinogen to trypsin. It recognizes a highly specific amino acid sequenc DDDDK and cleaves after lysine residues. The high specific activity of Enteropeptidase has been utilized in cleaving a variety of native or fusion proteins containing the above recognition motif. Biovisions Enteropeptidase Cleavage Kit contains highly active light chain fragment of human Enteropeptidase. Our pure enzyme is an excellent tool to obtain a wild type protein sequence from a fusion protein, containing Enteropeptidase recognition sequence (DDDDK). This Enteropeptidase cleavage kit is sufficient for cleaving at least 5 mg of target protein. The residual enteropeptidase left in the reaction mix will not interfere with most of the downstream applications. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9884