Learn More
Enteropeptidase/Enterokinase Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0259- 8S12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212411AF532
Catalog No. NP212411A32
Add to cart
Description
Description
Enteropeptidase/Enterokinase Monoclonal antibody specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot.
Specifications
Specifications
Enteropeptidase/Enterokinase
MM0259- 8S12
Western Blot
EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
TMPRSS15
Human recombinant Enteropeptidase (also called Enterokinase).
0.1 mL
Primary
5651