Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterovirus 71 3D Mouse anti-Virus, Unconjugated, Clone: 4, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen Enterovirus 71 3D
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF22351MI
View Documents
MilliporeSigma
MABF2235100UG
100 μg
Each for $372.00
Add to cart
 
MABF22352MI
View Documents
MilliporeSigma
MABF223525UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

Enterovirus 71 3D Monoclonal antibody specifically detects Enterovirus 71 3D in Virus samples. It is validated for Immunofluorescence, Immunoprecipitation, Western Blotting.
Specifications

Specifications

Enterovirus 71 3D
Unconjugated
Mouse
EV71;EV71 3D polymerase;Human rhinovirus A71
Recombinant 3D polymerase from Enterovirus 71.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunofluorescence, Immunoprecipitation, Western Blot
Purified
Virus
CN12_48981gpPO
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit