Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENTHD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ENTHD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170483
|
Novus Biologicals
NBP170483 |
100 μL |
Each for $436.00
|
|
NBP17048320
|
Novus Biologicals
NBP17048320UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
ENTHD1 Polyclonal specifically detects ENTHD1 in Human samples. It is validated for Western Blot.Specifications
ENTHD1 | |
Polyclonal | |
Rabbit | |
Human | |
ENTH domain containing 1 | |
ENTHD1 | |
IgG | |
Affinity Purified | |
67 kDa |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
150350 | |
Synthetic peptides corresponding to ENTHD1(ENTH domain containing 1) The peptide sequence was selected from the middle region of ENTHD1. Peptide sequence RAIARLHEDLSTVIQELNVINNILMSMSLNSSQISQSSQVPQSSEGSSDQ. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title