Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENTHD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144731
Description
Immunogen sequence: SECTQLRALC AIASLGSSDL LPQEHILLRT RPWLQELSMG SPGPVTNKAT KILRHFEASC GQLSPARGTS AEPGPTAALP GPSDLLTDAV PLPGSQVFLQ PLSSTPVSSR SPAPSSGMP Antigen sequence identity: Mouse 67%; Rat 69%.
ENTHD2 is a protein coding gene.
Specifications
ENTHD2 | |
Polyclonal | |
Unconjugated | |
TEPSIN | |
2410002I01Rik; AP-4 accessory protein; AP-4 complex accessory subunit tepsin; C17orf56; C19H17orf56; C9H17orf56; ENTH domain containing 2; ENTH domain-containing protein 2; Enthd2; Epsin for AP-4; protein C17orf56 homolog; RGD1307410; TEPSIN; TEPSIN adaptor related protein complex 4 accessory protein; TEPSIN, adaptor related protein complex 4 accessory protein; tetra-epsin | |
Rabbit | |
Affinity chromatography | |
RUO | |
146705 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96N21 | |
TEPSIN | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 293-411. Recombinant protein control fragment (Product #RP-110171). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction