Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENTPD1 Rabbit anti-Human, Polyclonal , Abnova™

ENTPD1 polyclonal antibody raised against recombinant human ENTPD1.

Manufacturer:  Abnova Corporation PAB29490

Catalog No. 89-934-090


Add to cart

Description

Description

Sequence: GVVHQVEECRVKGPGISKFVQKVNEIGIYLTDCMERAREVIPRSQHQETPVYLGATAGMRLLRMESEELADRVLDVVERSLSNYPFDFQGARIITGQEEGAYGWITINYLLGKFSQKTRWFS
Specifications

Specifications

ENTPD1
Unconjugated
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Liquid
ENTPD1
Recombinant protein corresponding to amino acids of human ENTPD1.
Antigen affinity purification
RUO
Primary
953
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
ENTPD1 polyclonal antibody raised against recombinant human ENTPD1.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
ATPDase/CD39/DKFZp686D194/DKFZp686I093/FLJ40921/FLJ40959/NTPDase-1
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit