Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENTPD4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548966

Catalog No. PA548966

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Hydrolyzes preferentially nucleoside 5 ft.-diphosphates, nucleoside 5 ft.-triphosphates are hydrolyzed only to a minor extent. The order of activity with different substrates is UDP >> GDP = CDP = TDP, AMP, ADP, ATP and UMP are not substrates. Preferred substrates for isoform 2 are CTP, UDP, CDP, GTP and GDP, while isoform 1 utilizes UTP and TTP.
Specifications

Specifications

ENTPD4
Polyclonal
PBS with 0.09% sodium azide
Q9Y227, Q9DBT4
Ectonucleoside triphosphate diphosphohydrolase 4, NTPDase 4, Lysosomal apyrase-like protein of 70 kDa, Uridine-diphosphatase, UDPase, ENTPD4, KIAA0392, LALP70, LYSAL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
67464, 9583
Western Blot
Unconjugated
ENTPD4
Liquid
ENTPD4
KLH conjugated synthetic peptide between 40-69 amino acids from the N-terminal region of human ENTPD4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit