Learn More
Thermo Scientific™ Entranceposons
Create transposons containing sequences of interest with these artificial DNA transposons available as ready-to-use linear transposons or replicable plasmids.
Specifications
Storage Requirements -20°C
View More Specs
Products 4
Catalog Number Mfr. No. Marker Gene Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Marker Gene Quantity Price Quantity & Availability  
F760
View Documents
Thermo Scientific™
F760
M1-CamR 10µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
F762
View Documents
Thermo Scientific™
F762
M1-KanR 10µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
F771
View Documents
Thermo Scientific™
F771
supF 20µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
F773
View Documents
Thermo Scientific™
F773
supF 40µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
Entranceposons are artificial DNA transposons. They are available as ready-to-use linear transposons or replicable plasmids. The plasmid Entranceposons allow the user to create transposons containing additional sequences of interest, e.g. marker genes. These custom transposons can then easily be inserted into the target DNA by taking advantage of the efficient Mu transposition reaction.
Entranceposons are artificial DNA transposons. They are available as ready-to-use linear transposons that can easily be inserted into the target DNA by taking advantage of the efficient Mu transposition reaction.
Specifications
Specifications
-20°C