Learn More
Thermo Scientific™ Entranceposons
Create transposons containing sequences of interest with these artificial DNA transposons available as ready-to-use linear transposons or replicable plasmids.
Manufacturer:  Thermo Scientific™ F773
 View more versions of this product
Catalog No. F773
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
Description
Description
Entranceposons are artificial DNA transposons. They are available as ready-to-use linear transposons or replicable plasmids. The plasmid Entranceposons allow the user to create transposons containing additional sequences of interest, e.g. marker genes. These custom transposons can then easily be inserted into the target DNA by taking advantage of the efficient Mu transposition reaction.
Entranceposons are artificial DNA transposons. They are available as ready-to-use linear transposons that can easily be inserted into the target DNA by taking advantage of the efficient Mu transposition reaction.
Specifications
Specifications
constructing custom Entranceposons
pEntranceposons
-20°C
supF
40µL