Learn More
Sartorius™ Entris™ 90 mm Analytical Balances
Provides speedy results for heavy workloads with repetitive applications. Sartorius™ Entris™ 90mm Analytical Balances feature a new operating concept: easy configuration of individual settings guided by short, menu-driven text prompts and simple navigation using cursor keys.
Products 6
Catalog Number Mfr. No. Capacity Model Stabilization Time Calibration Points Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Model Stabilization Time Calibration Points Price Quantity & Availability  
14-557-448PM
View Documents
Sartorius™
ENTRIS641SPM
60 g ENTRIS64-1S 2.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-447PM
View Documents
Sartorius™
ENTRIS1241SPM
120 g ENTRIS124-1S 2.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-446PM
View Documents
Sartorius™
ENTRIS2241SPM
220 g ENTRIS224-1S 2.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-016PM
View Documents
Sartorius™
ENTRIS64I1SPM
60 g ENTRIS64i-1S 2.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-017PM
View Documents
Sartorius™
ENTRIS124I1SPM
120 g ENTRIS124i-1S 2.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-018PM
View Documents
Sartorius™
ENTRIS224I1SPM
220 g ENTRIS224i-1S 2.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • Choice of 5 languages
  • External calibration only
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
  • Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting