Learn More
Sartorius™ Entris™ Precision Balances - PROMO
Delivers effective and reliable weighing results in daily work. Sartorius™ Entris™ Precision Balances provide easy and clearly structured user interface, logical key assignment and excellent readability for error-free operation.
Products 8
Catalog Number Mfr. No. Capacity Readability Model Stabilization Time Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Readability Model Stabilization Time Price Quantity & Availability  
14-557-452PM
View Documents
Sartorius™
ENTRIS1531SPM
1 mg ENTRIS153-1S 1.3 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-451PM
View Documents
Sartorius™
ENTRIS3231SPM
1 mg ENTRIS323-1S 1.1 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-450PM
View Documents
Sartorius™
ENTRIS4231SPM
1 mg ENTRIS423-1S 1.0 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-449PM
View Documents
Sartorius™
ENTRIS6231SPM
1 mg ENTRIS623-1S 1.0 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-019PM
View Documents
Sartorius™
ENTRIS153I1SPM
1 mg ENTRIS153i-1S 1.3 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-020PM
View Documents
Sartorius™
ENTRIS323I1SPM
1 mg ENTRIS323i-1S 1.1 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-021PM
View Documents
Sartorius™
ENTRIS423I1SPM
1 mg ENTRIS423i-1S 1.0 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-022PM
View Documents
Sartorius™
ENTRIS623I1SPM
1 mg ENTRIS623i-1S 1.0 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • Available with Internal or External calibration 
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting