Learn More
Sartorius™ Entris™ Toploading Balances - PROMO
When you need to get a heavy workload of repetitive applications done fast
Manufacturer:  Sartorius™ ENTRIS822I1SPM
 View more versions of this product
Catalog No. 14-559-023PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • External calibration only
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Specifications
Specifications
150mm
Toploading Balance
Not Applicable
Internal Calibration
10mg
1.5 sec.
820 g
RS-232C, 25-pin
10 mg
30mg
ENTRIS822i-1S
Not Applicable
Documents
Promotions
Promotions
  • Sartorius , Fisher Scientific , Fisherbrand
    Buy $1,500 or more in Sartorius products and get a Sartorius Tacta Pipette Multipack at no additional cost.
    06-30-2021