Learn More
Sartorius™ Entris™ Toploading Balances - PROMO
When you need to get a heavy workload of repetitive applications done fast
Specifications
Connection Type RS-232C, 25-pin
View More Specs
Products 16
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity & Availability  
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity & Availability  
14-557-457PM
View Documents
Sartorius™
ENTRIS8221SPM
ENTRIS822-1S 820 g 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-456PM
View Documents
Sartorius™
ENTRIS22021SPM
ENTRIS2202-1S 2.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-455PM
View Documents
Sartorius™
ENTRIS32021SPM
ENTRIS3202-1S 3.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-454PM
View Documents
Sartorius™
ENTRIS42021SPM
ENTRIS4202-1S 4.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-453PM
View Documents
Sartorius™
ENTRIS62021SPM
ENTRIS6202-1S 6.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-460PM
View Documents
Sartorius™
ENTRIS22011SPM
ENTRIS2201-1S 2.2 kg 100 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-459PM
View Documents
Sartorius™
ENTRIS52011SPM
ENTRIS5201-1S 5.2 kg 100 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-557-458PM
View Documents
Sartorius™
ENTRIS82011SPM
ENTRIS8201-1S 8.2 kg 100 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-023PM
View Documents
Sartorius™
ENTRIS822I1SPM
ENTRIS822i-1S 820 g 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-024PM
View Documents
Sartorius™
ENTRIS2202I1SPM
ENTRIS2202i-1S 2.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-025PM
View Documents
Sartorius™
ENTRIS3202I1SPM
ENTRIS3202i-1S 3.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-026PM
View Documents
Sartorius™
ENTRIS4202I1SPM
ENTRIS4202i-1S 4.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-027PM
View Documents
Sartorius™
ENTRIS6202I1SPM
ENTRIS6202i-1S 6.2 kg 10 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-028PM
View Documents
Sartorius™
ENTRIS2201I1SPM
ENTRIS2201i-1S 2.2 kg 100 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-029PM
View Documents
Sartorius™
ENTRIS5201I1SPM
ENTRIS5201i-1S 5.2 kg 100 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-559-030PM
View Documents
Sartorius™
ENTRIS8201I1SPM
ENTRIS8201i-1S 8.2 kg 100 mg 1.5 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • External calibration only
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applicaitons
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Specifications
Specifications
RS-232C, 25-pin