Learn More
Environmental Express™ Environmental Express™ TCLP Syringes
Manufacturer:  Environmental Express™ S1050
 View more versions of this product
Catalog No. 09-735-58
Add to cart
Specifications
Specifications
50mL
For volatile sample collection
TCLP syringe
Luer locking valve
Borosilicate Glass