Learn More
Environmental Slide Set 1: Environmental Pollution
See effects of environmental pollution in microscopic detail.
Supplier: Johannes Lieder GmbH & Co. KG 4510
Includes:
- Pine, healthy leaves
- Pine leaves damaged by acid rain
- Fir, healthy leaves
- Fir stem tip, damaged
- Beech, healthy leaves
- Beech leaves with destroyed epidermis and chloroplasts
- Tar spot of maples — consequence of single-crop farming
- Early leaf fall caused by thawing salt
- Healthy lichen
- Damaged lichen caused by air pollution
- Healthy wood of Beech
- Wood destroyed by fungus
- Wood rot fungus
- Root nodules with symbiotic bacteria
- Spruce beetle larva
- Wood with normal annual rings
- Wood with narrow annual rings caused by drought
- Bark with larval galleries of Spruce beetle
- Gall on Spruce caused by lice
- Gall on Oak caused by insects
Description
Set of 20 microscope slides demonstrates how environmental pollution affects plants and trees.
Specifications
Environment |