Learn More
Bioassay SystemsSupplier Diversity Partner ENZYCHROM FRUCTOSE ASSAY KIT DFS Item
Manufacturer:  Bioassay SystemsSupplier Diversity Partner EFRU100
Assay kit, fructose. Method: OD565nm. Samples: serum, plasma, urine, saliva, milk, culture medium, food, beverage, agriculture etc. Species: all. Procedure: 70 min. Size: 100 tests. Detection limit: 12 uM.
Catalog No. 50-489-269