Learn More
Bioassay Systems ENZYCHROM SUCROSE ASSAY KIT
SDP
Supplier:  Bioassay Systems ESUC100
Encompass_Preferred
Assay kit, sucrose. Method: OD565nm. Samples: serum, plasma, urine, saliva, milk, culture medium, food, beverage, agriculture etc. Species: all. Procedure: 70 min. Size: 100 tests. Detection limit: 17 uM.
Catalog No. 50-489-293
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.