Learn More
Bioworld Eosin Methylene Blue Agar (HOLT, Harris), 2 kg
Supplier: Bioworld 306200253
Eosin Methylene Blue Agar, Modified (formula of Holt-Harris and Teague)Slightly selective and differential medium for the isolation, cultivation and differentiation of gram-negative enteric bacillipH: 7.2Working Concentration: 36 g/LDehydrated Appearance: light red-purple, homogeneous powderPrepared Appearance: dark blue purple agarSoluble in boiling waterComposition:Pancreatic Digest of Gelatin Lactose Sucrose Dipotassium Phosphate Eosin Y Methylene Blue Agar
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.