Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosin Y Yellow Certified Macron Fine Chemicals™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass Bottle
17372-87-1
C20H12Br4Na2O8
745.904
MASXMTNVNASWNH-UHFFFAOYSA-L
2,4,5,7-tetrabromogluorescein
91886399
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
17372-87-1
C20H12Br4Na2O8
745.904
MASXMTNVNASWNH-UHFFFAOYSA-L
2,4,5,7-tetrabromogluorescein
91886399
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]
Formula Weight 691.86
Packaging Glass Bottle
Quantity 25g
Chemical Name or Material Eosin Y

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.