Learn More
Athens Research And Technology IncSupplier Diversity Partner EOSINOPHIL DN HU 100UG DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 1615050414100UG
Eosinophil Derived Neurotoxin, Human Eosinophils (EDN): 100 UG; Frozen in 5 mM NaH2PO4, pH 7.0, and 0.2 M NaCI; MW 18,000; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from whole blood in the Athens Research & Technology laboratory. Molecular Biology Reagent
Catalog No. 50-489-735