Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eotaxin Mouse anti-Human, Clone: 13H2, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $450.00
Specifications
Antigen | Eotaxin |
---|---|
Clone | 13H2 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC170425
|
MilliporeSigma
MABC170425UG |
25 μg |
Each for $177.00
|
|
|||||
MABC1704100
|
MilliporeSigma
MABC1704100UG |
100 μg |
Each for $450.00
|
|
|||||
Description
Eotaxin Monoclonal antibody specifically detects Eotaxin in Human samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).Specifications
Eotaxin | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
13H2 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Full-length recombinant mature human Eotaxin | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title