Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ ep Dualfilter T.I.P.S.™ 384 Filter Tips

Eppendorf™ ep Dualfilter T.I.P.S.™ 384 Filter Tips utilize a unique two-phase filter that provides ultimate sample and pipette protection

Specifications

Certifications/Compliance EN 1822
Color Clear
Filtered Filtered
For Use With (Equipment) Air-Cushion
Format Rack
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Volume (Metric) Price Quantity & Availability  
05-414-041
View Documents
Eppendorf™
0030078853
42 mm 0.1 to 20 μL N/A
05-414-042
View Documents
Eppendorf™
0030078861
53 mm 0.5 to 100 μL N/A
Description

Description

  • With a unique two-phase filter, ep Dualfilter T.I.P.S. filter tips offer the ultimate protection of pipette and sample against unintentional and unnoticed contamination
  • With defined flow dynamics, low wettability, and high thermal stability they are certified pyrogen-free and PCR clean.
  • 384 pipette tips build a system together with Research plus and Xplorer plus 16 and 24-channel pipettes for utmost safety and reliability
  • Innovative SOFTattach technology allows for reliable tip fit and tightness while operating forces are reduced to a minimum
  • Fill a whole 384-well plate without loosening pipette tips
  • A perfect tip alignment is guaranteed by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of ep Dualfilter T.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Dual protection for pipette and sample against aerosols, and biomolecules
  • Lot-specific certificates and certificates of filter efficiency acc. to EN 1822 are available
  • Available as racks and in purity grades PCR clean and sterile
  • Modern slim rack design with reclosable lid
  • Improved sustainability with 20% less polypropylene consumption per rack
Specifications

Specifications

EN 1822
Filtered
Rack
Sterile
3840 tips
Clear
Air-Cushion
Consumables
10 racks × 384 tips
PCR Clean
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit