Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ 384, PCR Clean and Sterile
ep Dualfilter T.I.P.S. 384 with the innovative SOFTattach technology are optimally coordinated to the Eppendorf 16 and 24-channel pipettes Research plus and Xplorer plus. Process 384-well plates manually with highest level of filter pipette tip tightness and coaxiality but extraordinary low operating forces.
Manufacturer:  Eppendorf™ 0030078403
 View more versions of this product
Catalog No. 05-413-613
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • ep Dualfilter T.I.P.S. 384 pipette tips build a system together with Research plus and Xplorer plus 16 and 24-channel pipettes for utmost safety and reliability
  • Innovative SOFTattach technology allows for reliable tip fit and tightness while operating forces are reduced to a minimum
  • Fill a whole 384-well plate without loosening pipette tips
  • A perfect tip alignment is guaranteed by extraordinary coaxiality for safe plate maneuvering and liquid transfer
  • The tip shape of ep Dualfilter T.I.P.S. 384 is adjusted to perfectly fit the wells of 384-well plates
  • Dual protection for pipette and sample against aerosols and biomolecules
  • Lot-specific certificates are available at: (URL: https://www.eppendorf.com/certificates)[certificates homepage].
  • Certificate of filter efficiency acc. to EN 1822 available at: (URL: https://www.eppendorf.com/certificates)[certificates homepage]
  • Available as racks
  • Available in purity grades of: PCR clean and sterile
  • 1) An aerosol is a colloid of fine solid particles or liquid droplets in air or another gas.
Specifications
Specifications
Yes
Polypropylene
Non-filtered
Clear
ep Dualfilter T.I.P.S.™ 384
PCR clean and sterile, Forensic DNA Grade, Eppendorf Quality™
Rack
10 racks × 384 tips
Pearl White
epT.I.P.S.™
20 to 0.1 μL
Sterile
Not Autoclavable
42 mm
Room temperature
Universal
3,840 tips