Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ ep Dualfilter T.I.P.S.™ Forensic DNA Grade Filter Tips

Eppendorf™ ep Dualfilter T.I.P.S.™ Forensic DNA Grade Filter Tips offer ultimate sample protection, are certified pyrogen-free, and PCR clean

$205.50 - $219.50

Specifications

Certifications/Compliance EN 1822
Color Clear
Filtered Filtered
For Use With (Equipment) Air-Cushion
Format Rack
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
05-413-971
View Documents
Eppendorf™
0030078810
0.1 to 10 μL
Pack of 960 for $219.50
Add to cart
 
05-413-972
View Documents
Eppendorf™
0030078829
2 to 20 μL
Pack of 960 for $219.50
Add to cart
 
05-413-973
View Documents
Eppendorf™
0030078837
2 to 200 μL
Pack of 960 for $205.50
Add to cart
 
05-413-974
View Documents
Eppendorf™
0030078845
50 to 1000 μL
Pack of 960 for $219.50
Add to cart
 
Description

Description

  • With a unique two-phase filter, ep Dualfilter T.I.P.S. filter tips offer the ultimate protection of pipette and sample against unintentional and unnoticed contamination
  • With defined flow dynamics, low wettability, and high thermal stability they are certified pyrogen-free and PCR clean
  • Two filter layers provide dual protection against aerosols and biomolecules to guard pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Certificate of filter efficiency according to EN 1822 is also available
  • Modern slim rack design with reclosable lid
  • Improved sustainability with 20% less polypropylene consumed per rack
Specifications

Specifications

EN 1822
Filtered
Rack
10 racks × 96 tips
Clear
Air-Cushion
Consumables
960 tips
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit