Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ LoRetention Tips
Eppendorf™ ep Dualfilter T.I.P.S.™ LoRetention Tips have an ultrahomogeneous, and ultrahydrophobic surface for maximum retention and reproducibility
Manufacturer:  Eppendorf™ 0030078640
 View more versions of this product
Catalog No. 05-413-960
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Designed for maximum recovery and reproducibility, ep Dualfilter T.I.P.S. LoRetention tips have an ultrahomogeneous and ultrahydrophobic surface
  • Significantly reduced foam formation during pipetting
  • No sample loss, reduced costs, and improved results
  • Ultrahomogenous surface for maximized reproducibility
  • Ultrahydrophobic surface for minimal sample loss
  • Extreme resistance to chemicals
  • Available in PCR clean and Eppendorf Quality
  • No coating
  • Modern slim rack design with reclosable lid
  • Improved sustainability with 20% less polypropylene consumed per rack
Specifications
Specifications
Air-Cushion
Filtered
Clear
46 mm
PCR Clean
Rack
Consumables
Light Gray
0.5 to 20 μL
Sterile
LoRetention
Lot-Specific Certificates Available
Replaces Order No. 0030077628
960 tips
10 racks × 96 tips