Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ ep Dualfilter T.I.P.S.™ LoRetention Tips

Eppendorf™ ep Dualfilter T.I.P.S.™ LoRetention Tips have an ultrahomogeneous, and ultrahydrophobic surface for maximum retention and reproducibility

$199.60 - $217.50

Specifications

Certifications/Compliance Lot-Specific Certificates Available
Color Clear
Filtered Filtered
For Use With (Equipment) Air-Cushion
Format Rack
View More Specs

Products 6
Catalog Number Mfr. No. Length (Metric) Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Volume (Metric) Price Quantity & Availability  
05-413-960
View Documents
Eppendorf™
0030078640
46 mm 0.5 to 20 μL
Pack of 960 for $217.50
Add to cart
 
05-413-961
View Documents
Eppendorf™
0030078659
53 mm 2 to 100 μL
Pack of 960 for $199.60
Add to cart
 
05-413-962
View Documents
Eppendorf™
0030078667
55 mm 2 to 200 μL
Pack of 960 for $202.00
Add to cart
 
05-413-964
View Documents
Eppendorf™
0030078683
76 mm 50 to 1000 μL
Pack of 960 for $217.50
Add to cart
 
05-413-959
View Documents
Eppendorf™
0030078632
34 mm 0.1 to 10 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
05-413-963
View Documents
Eppendorf™
0030078675
55 mm 20 to 300 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Designed for maximum recovery and reproducibility, ep Dualfilter T.I.P.S. LoRetention tips have an ultrahomogeneous and ultrahydrophobic surface
  • Significantly reduced foam formation during pipetting
  • No sample loss, reduced costs, and improved results
  • Ultrahomogenous surface for maximized reproducibility
  • Ultrahydrophobic surface for minimal sample loss
  • Extreme resistance to chemicals
  • Available in PCR clean and Eppendorf Quality
  • No coating
  • Modern slim rack design with reclosable lid
  • Improved sustainability with 20% less polypropylene consumed per rack
Specifications

Specifications

Lot-Specific Certificates Available
Filtered
Rack
Sterile
10 racks × 96 tips
LoRetention
Clear
Air-Cushion
Consumables
PCR Clean
960 tips
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit