Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epac2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282089JF646
This item is not returnable.
View return policy
Description
Epac2 Polyclonal antibody specifically detects Epac2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Epac2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
CAMP-GEFII, cAMP-regulated guanine nucleotide exchange factor II, CGEF2EPAC 2, EPAC2, Exchange factor directly activated by cAMP 2, Exchange protein directly activated by cAMP 2, Nbla00496, putative protein product of Nbla00496, Rap guanine nucleotide exchange factor (GEF) 4, rap guanine nucleotide exchange factor 4, RAP guanine-nucleotide-exchange factor (GEF) 4 | |
EPAC2 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human EPAC2. The immunogen is located within the first 50 amino acids of EPAC2. Amino Acid Squence: LDKRPLERSSEDVD | |
0.1 mL | |
Signal Transduction | |
11069 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction