Learn More
Aviva Systems Biology Corp EPB41L2 Antibody - middle region (ARP54569_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54569_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EPB41L2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EPB41L2. Alternate names: RP3-324N14.1, 4.1-G, DKFZp781D1972, DKFZp781H1755, 4.1GApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61244