Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPB41L4A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548831

Catalog No. PA548831

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Members of the band 4.1 protein superfamily, including EPB41L4A, are thought to regulate the interaction between the cytoskeleton and plasma membrane (Ishiguro et al., 2000 [PubMed 10874211]).[supplied by OMIM, Jul 2008]
Specifications

Specifications

EPB41L4A
Polyclonal
PBS with 0.09% sodium azide
Q9HCS5
Band 41-like protein 4A, Protein NBL4, EPB41L4A, EPB41L4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64097
Western Blot
Unconjugated
EPB41L4A
Liquid
EPB41L4A
KLH conjugated synthetic peptide between 562-590 amino acids from the C-terminal region of human EPB41L4A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit