Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525141
Description
This antibody is predicted to react with Xenopus based on sequence homology.
EPC2 may play a role in transcription or DNA repair.
Specifications
EPC2 | |
Polyclonal | |
Unconjugated | |
EPC2 | |
5830499L14; D2Ertd694e; Enhancer of polycomb homolog 2; enhancer of polycomb homolog 2 (Drosophila); Epc2; EPC-LIKE | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
227867, 26122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q52LR7, Q8C0I4 | |
EPC2 | |
KLH conjugated synthetic peptide between 205-233 amino acids from the N-terminal region of human EPC2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction