Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpCAM Rabbit anti-Human, Unconjugated, Clone: 1B8, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen EpCAM
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB121525MI
View Documents
MilliporeSigma
ZRB121525UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12154XMI
View Documents
MilliporeSigma
ZRB12154X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

EpCAM Monoclonal antibody specifically detects EpCAM in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

EpCAM
Unconjugated
Rabbit
Epithelial cell adhesion molecule;Adenocarcinoma-associated antigen;Cell surface glycoprotein Trop-1;Epithelial cell surface antigen;Epithelial glycoprotein;EGP;Epithelial glycoprotein 314;EGP314;hEGP314;KS 1/4 antigen;KSA;Major gastrointestinal tumor-associated protein GA733-2;Tumor-associated calcium signal transducer 1;CD326
GST/His-tagged recombinant fragment corresponding to 140 amino acids from the extracellular domain of human Epithelial cell adhesion molecule (EpCAM).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human
EPCAM, GA733-2, M1S2, M4S1, MIC18, TACSTD1, TROP1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit