Learn More
Novus Biologicals™ EpCAM/TROP1 Antibody Pair [HRP]
Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2795125PLATES
 View more versions of this product
Catalog No. NB010064
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human EpCAM/TROP1.
Specifications
Specifications
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human EpCAM/TROP1.
15.625 pg/mL
4072
RUO
EpCAM/TROP1
Cancer, Cancer Stem Cells
Human
EPCAM
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended.
HRP
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 180 ng of recombinant Human EpCAM/TROP1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Human EpCAM/TROP1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human EpCAM/TROP1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    15.625-1000 pg/mL
    17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A