Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EpCAM/TROP1 Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793785PLATES

 View more versions of this product

Catalog No. NB009796


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse EpCAM/TROP1.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
78 pg/mL
4072
RUO
EpCAM/TROP1
Cancer, Cancer Stem Cells
Mouse
EPCAM
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended.
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse EpCAM/TROP1.
Sandwich ELISA
  • Standard: Each vial contains 400 ng of recombinant mouse EpCAM/TROP1.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-mouse EpCAM/TROP1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-EpCAM/TROP1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Storage is content dependent.
    Rabbit
    5 Plates
    78.13-5000 pg/mL
    17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit