Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human EpCAM/TROP1 His Protein
SDP

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$398.00 - $1472.00

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 4072
Molecular Weight (g/mol) TMW: 28.2kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB252195002
SDP
View Documents
Novus Biologicals™
NBP2521900.05MG
0.05mg
Each for $398.00
Add to Cart
 
NBP25219025
SDP
View Documents
Novus Biologicals™
NBP2521900.25MG
0.25mg
Each for $1,472.00
Add to Cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 24-265 of Human EpCAM/TROP1 The Recombinant Human EpCAM/TROP1 Protein is derived from Baculovirus. The Recombinant Human EpCAM/TROP1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

1mg/mL
Phosphate buffered saline (pH 7.4) containing 10% glycerol
TMW: 28.2kDa
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
EpCAM/TROP1
Recombinant
SDS-PAGE
4072
Baculovirus
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
SDS
Documents

Documents

Product Certifications

For research use only and is not approved for use in humans or in clinical diagnosis

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit