Learn More
EpCAM/TROP1 Mouse anti-Human, Alexa Fluor 405, Clone: 158206, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB9601V100UG
Catalog No. FAB9601V100
Add to cart
This item is not returnable. View return policy
Description
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
EpCAM/TROP1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Mouse
IgG2b
RUO
Primary
4072
Human
Flow Cytometry
158206
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
EPCAM
Mouse myeloma cell line NS0-derived recombinant human EpCAM/TROP-1 Extracellular domain
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human EpCAM/TROP-1 in ELISAs.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)